Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013912.3 | NP_038940.1 | 2 | 448 | ||
GenBank mRNA | AB023494.1 | - | 2 | 101 | ||
AJ290423.1 | - | 2 | 200 | |||
AK042076.1 | - | 2 | 452 | |||
BB618863.1 | - | 2 | 448 | |||
BC020015.1 | - | 2 | 429 | 3483588 | ||
DQ832282.1 | - | 2 | 437 |