Search
Search

Zfp653| Gene Symbol: Zfp653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177318.2 | NP_796292.1 | 6 | 1433 | ||
| GenBank mRNA | AK046492.1 | - | 5 | 1480 | ||
| AK089104.1 | - | 6 | 1245 | |||
| AK157635.1 | - | 5 | 1480 | |||
| AY072705.1 | - | 6 | 1443 | |||
| BC038037.1 | - | 6 | 1433 | 4190337 | ||