Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gchfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177157.4 | NP_796131.1 | 2 | 161 | ||
GenBank mRNA | AK028116.1 | - | 2 | 162 | ||
AK028122.1 | - | 2 | 162 | |||
BC096406.1 | - | 2 | 149 | 1511037 | ||
BC132450.1 | - | 2 | 157 | 40130727 | ||
BC132478.1 | - | 2 | 157 | 40130755 |