Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194348.3 | NP_919329.2 | 8 | 1290 | ||
GenBank mRNA | AK129132.1 | - | 8 | 1215 | ||
BC023754.1 | - | 4 | 572 | |||
BC063264.1 | - | 8 | 1231 | 6409527 |