Search
Search

Hecw2| Gene Symbol: Hecw2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001883.3 | NP_001001883.1 | 5 | 608 | ||
| NM_172655.3 | NP_766243.2 | 5 | 608 | |||
| GenBank mRNA | AB182244.1 | - | 4 | 514 | ||
| AK042922.1 | - | 5 | 609 | |||
| AK129325.1 | - | 3 | 145 | |||
| BC107219.2 | - | 5 | 593 | |||
| BC134353.1 | - | 5 | 592 | |||
| BC148188.2 | - | 5 | 591 | |||