Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177845.4 | NP_808513.2 | 19 | 2792 | ||
GenBank mRNA | AB195277.1 | - | 19 | 2695 | ||
BC060600.1 | - | 16 | 2738 | |||
BC066085.1 | - | 19 | 2792 | |||
BC137899.1 | - | 19 | 2569 | 8860918 | ||
BC145020.1 | - | 20 | 2680 |