Search Thermo Fisher Scientific
Gene Symbol: Mamld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081354.2 | NP_001074823.2 | 3 | 282 | ||
NM_001256048.1 | NP_001242977.1 | 3 | 282 | |||
GenBank mRNA | AK090190.1 | - | 3 | 283 | ||
BC157923.1 | - | 2 | 192 | 9088096 | ||
BC157989.1 | - | 2 | 192 | 9007403 | ||
BC172017.1 | - | 2 | 192 | 9054303 | ||
BY733469.1 | - | 3 | 283 |