Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033474.2 | NP_001028646.2 | 1 | 577 | ||
GenBank mRNA | AK003631.1 | - | 1 | 547 | ||
AK011696.1 | - | 1 | 551 | |||
AK014824.1 | - | 1 | 473 | |||
AK135540.1 | - | 1 | 550 | |||
AK150316.1 | - | 1 | 547 | |||
BC030168.1 | - | 1 | 546 | |||
BC047210.1 | - | 1 | 547 | |||
BC058561.1 | - | 1 | 509 | |||
BC079579.1 | - | 1 | 532 | |||
BC094390.1 | - | 1 | 535 |