Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RRM2B SIRNA Assays ›
Gene Symbol:
Rrm2b
Gene Name:
ribonucleotide reductase M2 B (TP53 inducible)
Gene Aliases:
p53R2
Chromosome Location:
Chr.15: 37923952 - 37961108 on Build GRCm38
UniGene ID:
Mm.24738
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS283985
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rrm2b
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_199476.1
NP_955770.1
5
694
GenBank mRNA
AK138731.1
-
5
687
BC058103.1
-
5
694
6827753
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001130543.
Panther Classification:
Molecular Function -
reductase
Gene Ontology Categories:
Function(s)
Process(es)
kidney development
renal system process
DNA replication
mitochondrial DNA replication
DNA repair
cellular response to DNA damage stimulus
response to oxidative stress
deoxyribonucleoside diphosphate metabolic process
deoxyribonucleoside triphosphate metabolic process
deoxyribonucleotide biosynthetic process
oxidation-reduction process
negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator
ribonucleoside-diphosphate reductase activity, thioredoxin disulfide as acceptor
oxidoreductase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0