Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114637.1 | NP_001108109.1 | 2 | 233 | ||
GenBank mRNA | BC016255.1 | - | 2 | 210 |
Gene Symbol: Gm28042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB195274.1 | - | 2 | 223 | ||
AB195275.1 | - | 2 | 223 |