Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135198.1 | NP_001128670.1 | 2 | 136 | ||
GenBank mRNA | BC058624.1 | - | 2 | 141 |