Search
Search

Il27ra| Gene Symbol: Il27ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016671.3 | NP_057880.2 | 5 | 702 | ||
| GenBank mRNA | AF053005.1 | - | 5 | 670 | ||
| AF265243.1 | - | 5 | 681 | |||
| AK040457.1 | - | 3 | 808 | |||
| AK133753.1 | - | 5 | 717 | |||
| BC032878.1 | - | 5 | 702 | 1244998 | ||