Search Thermo Fisher Scientific
Gene Symbol: Uevld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040695.1 | NP_001035785.1 | 8 | 954 | ||
GenBank mRNA | AK154536.1 | - | 8 | 905 | ||
AK155685.1 | - | 8 | 901 | |||
BC054796.1 | - | 8 | 934 | 6843964 |