Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Praf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138602.4 | NP_613068.1 | 1, 2 | 185 | ||
GenBank mRNA | AF229636.1 | - | 1, 2 | 172 | ||
AK003344.1 | - | 1, 2 | 186 | |||
AK224918.1 | - | 1, 2 | 186 | |||
BC064757.1 | - | 1, 2 | 210 | 30056020 |