Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019547.2 | NP_062420.2 | 4 | 693 | ||
GenBank mRNA | AI573584.1 | - | 4 | 712 | ||
AK052886.1 | - | 2 | 478 | |||
AK146026.1 | - | 4 | 667 | |||
AK162141.1 | - | 4 | 666 | |||
AK172625.1 | - | 4 | 664 | |||
BC006687.1 | - | 3 | 1202 | |||
BC085307.1 | - | 4 | 637 | 6396103 |