Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252463.1 | NP_001239392.1 | 4 | 307 | ||
NM_001252464.1 | NP_001239393.1 | 4 | 282 | |||
NM_001252465.1 | NP_001239394.1 | 2 | 91 | |||
NM_019988.5 | NP_064372.2 | 4 | 302 | |||
GenBank mRNA | AF237676.1 | - | 4 | 233 | ||
AK002751.1 | - | 4 | 268 | |||
AK038515.1 | - | 3 | 800 | |||
AK040100.1 | - | 4 | 303 | |||
AK048102.1 | - | 4 | 266 | |||
AK077680.1 | - | 4 | 273 | |||
AK083731.1 | - | 4 | 243 | |||
AK088951.1 | - | 4 | 276 | |||
BC015279.1 | - | 4 | 261 | 4222934 | ||
BM947104.1 | - | 2 | 73 | |||
CF750475.1 | - | 4 | 271 |