Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018758.2 | NP_061228.1 | 2 | 451 | ||
GenBank mRNA | AF070975.1 | - | 2 | 405 | ||
AK045965.1 | - | 2 | 452 | |||
AK082046.1 | - | 371 | ||||
AK169462.1 | - | 2 | 437 | |||
AK170182.1 | - | 2 | 399 | |||
BC005423.1 | - | 2 | 383 | 3487448 | ||
BC009666.1 | - | 2 | 395 | 3490710 |