Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GHRL SIRNA Assays ›
          Gene Symbol:
          Ghrl
          Gene Name:
          ghrelin
          Gene Aliases:
          2210006E23Rik, Ghr, MTLRP, MTLRPAP, m46
          Chromosome Location:
          Chr.6: 113716119 - 113727485 on Build GRCm38
          UniGene ID:
          Mm.379095
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS286178
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ghrl
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286404.1 NP_001273333.1 2, 3 254
          NM_021488.5 NP_067463.2 3, 4 305
          GenBank mRNA AB035701.1 - 2, 3 247
          AJ243503.1 - 2, 3 270
          AK008658.1 - 3, 4 275
          AK008860.1 - 3, 4 275
          BC132230.1 - 2, 3 271 40130507
          BC132232.1 - 2, 3 271 40130509
          FJ914266.1 - 2, 3 311

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_021669, AB029433, AB035699, BF543543.

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          gastric acid secretion
          negative regulation of endothelial cell proliferation
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          actin polymerization or depolymerization
          adult feeding behavior
          visual learning
          response to hormone
          hormone-mediated signaling pathway
          dendrite development
          positive regulation of insulin secretion
          regulation of response to food
          positive regulation of response to food
          positive regulation of appetite
          negative regulation of interleukin-1 beta production
          negative regulation of locomotion
          regulation of cell proliferation
          negative regulation of circadian sleep/wake cycle, REM sleep
          negative regulation of tumor necrosis factor biosynthetic process
          negative regulation of apoptotic process
          response to estrogen
          negative regulation of interleukin-6 biosynthetic process
          positive regulation of circadian sleep/wake cycle, non-REM sleep
          negative regulation of insulin secretion
          decidualization
          negative regulation of inflammatory response
          positive regulation of corticotropin secretion
          positive regulation of cortisol secretion
          positive regulation of synapse assembly
          excitatory postsynaptic potential
          positive regulation of growth hormone secretion
          positive regulation of protein tyrosine kinase activity
          negative regulation of energy homeostasis
          positive regulation of energy homeostasis
          G-protein coupled receptor binding
          hormone activity
          growth hormone-releasing hormone activity
          protein tyrosine kinase activator activity
          ghrelin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0