Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GSK3A SIRNA Assays ›
Gene Symbol:
Gsk3a
Gene Name:
glycogen synthase kinase 3 alpha
Gene Aliases:
2700086H06Rik
Chromosome Location:
Chr.7: 25228258 - 25237851 on Build GRCm38
UniGene ID:
Mm.491101
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS286307
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Gsk3a
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031667.1
NP_001026837.1
2
557
GenBank mRNA
BQ885209.1
-
2
150
CK624130.1
-
2
557
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of systemic arterial blood pressure
cardiac left ventricle morphogenesis
carbohydrate metabolic process
glycogen metabolic process
regulation of gene expression by genetic imprinting
protein phosphorylation
nervous system development
insulin receptor signaling pathway
negative regulation of signal transduction
positive regulation of peptidyl-threonine phosphorylation
Wnt signaling pathway
phosphorylation
cell migration
positive regulation of cAMP biosynthetic process
negative regulation of TOR signaling
cellular response to insulin stimulus
positive regulation of peptidyl-serine phosphorylation
cellular response to interleukin-3
proteasome-mediated ubiquitin-dependent protein catabolic process
hypermethylation of CpG island
positive regulation of heart contraction
positive regulation of transcription from RNA polymerase II promoter
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
negative regulation of transferase activity
negative regulation of cell growth involved in cardiac muscle cell development
cellular response to lithium ion
cellular response to organic cyclic compound
positive regulation of adrenergic receptor signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
regulation of mitophagy
positive regulation of protein targeting to mitochondrion
positive regulation of glycogen (starch) synthase activity
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase A catalytic subunit binding
tau-protein kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0