Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037916.3 | NP_001033005.1 | 8, 9 | 1004 | ||
GenBank mRNA | AK029257.1 | - | 8, 9 | 1006 | ||
AK043689.1 | - | 2, 3 | 324 | |||
AK076542.1 | - | 7, 8 | 975 | |||
BC150874.1 | - | 6, 7 | 791 | 9087785 | ||
BC172023.1 | - | 6, 7 | 791 | 9054309 |