Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252591.2 | NP_001239520.1 | 4 | 550 | ||
GenBank mRNA | AA434782.1 | - | 4 | 404 | ||
AF364070.1 | - | 4 | 389 | |||
AI035961.1 | - | 4 | 364 | |||
AJ245772.1 | - | 4 | 405 | |||
AK003105.1 | - | 4 | 397 | |||
AK009857.1 | - | 4 | 391 | |||
AK010172.1 | - | 4 | 397 | |||
AY026524.1 | - | 4 | 396 | |||
BC051197.1 | - | 4 | 349 | 1397202 | ||
BC061051.1 | - | 4 | 311 | 30300606 | ||
BC132358.1 | - | 4 | 225 | 40130635 | ||
BC138801.1 | - | 4 | 225 | 8861823 | ||
BQ176246.1 | - | 3 | 552 | |||
W18646.1 | - | 5 | 303 |