Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc28b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025455.2 | NP_079731.2 | 5, 6 | 668 | ||
GenBank mRNA | AK007427.1 | - | 4, 5 | 445 | ||
AK009176.1 | - | 3, 4 | 288 | |||
AK028262.1 | - | 5, 6 | 646 | |||
AK158016.1 | - | 5, 6 | 672 | |||
BC028873.1 | - | 5, 6 | 660 | 4220355 | ||
BQ031123.1 | - | 1, 2 | 185 |