Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024181.2 | NP_077143.2 | 9, 10 | 1335 | ||
GenBank mRNA | AF255459.1 | - | 9, 10 | 1012 | ||
AF314002.1 | - | 8, 9 | 848 | |||
AK004617.1 | - | 9, 10 | 1090 | |||
AK028184.1 | - | 9, 10 | 1075 | |||
AK128945.1 | - | 9, 10 | 1089 | |||
AK165652.1 | - | 9, 10 | 1094 | |||
AK169020.1 | - | 9, 10 | 1102 |