Search
Search

Tmem107| Gene Symbol: Tmem107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025838.2 | NP_080114.1 | 5 | 521 | ||
| NM_028336.3 | NP_082612.2 | 4 | 747 | |||
| GenBank mRNA | AK002804.1 | - | 5 | 491 | ||
| AK003403.1 | - | 5 | 517 | |||
| AK012938.1 | - | 4 | 746 | |||
| AK154279.1 | - | 5 | 519 | |||
| BC096686.1 | - | 5 | 499 | 1446796 | ||
| CO044635.1 | - | 1 | 532 | |||