Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038635.2 | NP_001033724.2 | 2 | 819 | ||
NM_183262.3 | NP_899085.3 | 2 | 819 | |||
GenBank mRNA | AK006778.1 | - | 2 | 584 | ||
AK165988.1 | - | 2 | 714 | |||
BC047277.1 | - | 1 | 339 | |||
BC138904.1 | - | 2 | 726 | 8861926 | ||
BC138905.1 | - | 2 | 723 | 8861927 |