Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026155.3 | NP_080431.1 | 4 | 395 | ||
GenBank mRNA | AK002818.1 | - | 4 | 396 | ||
AK029353.1 | - | 4 | 396 | |||
AK076235.1 | - | 4 | 428 | |||
AK088555.1 | - | 4 | 400 | |||
AK150255.1 | - | 4 | 389 | |||
AK162235.1 | - | 2 | 99 | |||
BC011111.1 | - | 4 | 402 | 3965897 | ||
BC020335.1 | - | 4 | 367 | |||
CT010172.1 | - | 4 | 379 |