Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026184.2 | NP_080460.2 | 8 | 834 | ||
GenBank mRNA | AF489856.1 | - | 8 | 640 | ||
AK004981.1 | - | 811 | ||||
AK006886.1 | - | 8 | 697 | |||
AK041951.1 | - | 8 | 810 | |||
AK160664.1 | - | 8 | 850 | |||
BC058721.1 | - | 8 | 834 | 5705507 | ||
BC111900.1 | - | 8 | 804 | 40062137 | ||
BC112426.1 | - | 8 | 804 | 40062136 | ||
BC145888.1 | - | 8 | 785 | 40131191 | ||
BC145890.1 | - | 8 | 785 | 40131193 |