Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc39d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026351.2 | NP_080627.2 | 1 | 1169 | ||
GenBank mRNA | AK016184.1 | - | 1 | 752 | ||
AK016869.1 | - | 1 | 616 | |||
BC118946.1 | - | 1 | 1169 | 40092366 | ||
BC118947.1 | - | 1 | 1169 | 40092368 |