Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dalrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026378.3 | NP_080654.3 | 2 | 390 | ||
GenBank mRNA | AK002439.1 | - | 2 | 387 | ||
AK149963.1 | - | 2 | 391 | |||
AK169405.1 | - | 2 | 388 | |||
AK171818.1 | - | 2 | 391 | |||
BC013483.1 | - | 2 | 371 | 4242135 |