Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncaip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199151.1 | NP_001186080.1 | 7 | 1567 | ||
NM_001199153.1 | NP_001186082.1 | 7 | 1472 | |||
NM_001199154.1 | NP_001186083.1 | 7 | 1742 | |||
NM_026408.4 | NP_080684.3 | 7 | 1567 | |||
GenBank mRNA | AF348447.1 | - | 7 | 1495 | ||
AK013040.1 | - | 5 | 1301 | |||
AK017012.1 | - | 4 | 502 | |||
AK044974.1 | - | 7 | 1765 | |||
AK081942.1 | - | 7 | 1746 | |||
AK082079.1 | - | 7 | 1571 | |||
AK132346.1 | - | 7 | 1670 | |||
AK160111.1 | - | 7 | 1571 | |||
BC098489.1 | - | 7 | 1471 | 30096466 | ||
BC130225.1 | - | 7 | 1457 | |||
BC152544.1 | - | 7 | 1455 |