Search
Search

Dynll2| Gene Symbol: Dynll2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168471.1 | NP_001161943.1 | 3 | 304 | ||
| NM_001168472.1 | NP_001161944.1 | 3 | 477 | |||
| NM_026556.4 | NP_080832.1 | 3 | 510 | |||
| GenBank mRNA | AK006876.1 | - | 3 | 286 | ||
| AK011284.1 | - | 3 | 478 | |||
| AK028319.1 | - | 3 | 275 | |||
| AK049645.1 | - | 3 | 484 | |||
| AK078434.1 | - | 3 | 483 | |||
| AK160114.1 | - | 3 | 480 | |||
| AK169212.1 | - | 3 | 483 | |||
| AY029255.1 | - | 2 | 212 | |||
| BC011289.1 | - | 3 | 301 | 3981362 | ||
| BC040822.1 | - | 3 | 472 | 4459858 | ||
| BC049621.1 | - | 283 | ||||