Search
Search

| Gene Symbol: Gtsf1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026630.2 | NP_080906.1 | 2 | 367 | ||
| GenBank mRNA | AK018886.1 | - | 1 | 307 | ||
| AK019131.1 | - | 2 | 368 | |||
| AK160150.1 | - | 1 | 267 | |||
| BC048439.1 | - | 1 | 240 | 6774366 | ||
| Gene Symbol: Zfp850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254951.1 | NP_001241880.1 | 6 | 5632 | ||
| GenBank mRNA | AK081702.1 | - | 1 | 2965 | ||
| BC025082.1 | - | 1 | 412 | 2648621 | ||