Search
Search

Gtsf1l| Gene Symbol: Gtsf1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026630.2 | NP_080906.1 | 2 | 498 | ||
| GenBank mRNA | AK018886.1 | - | 1 | 438 | ||
| AK019131.1 | - | 2 | 499 | |||
| AK160150.1 | - | 1 | 398 | |||
| BC048439.1 | - | 1 | 371 | 6774366 | ||