Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039188.1 | NP_001034277.1 | 10 | 1912 | ||
NM_001177868.1 | NP_001171339.1 | 10 | 1637 | |||
NM_001177869.1 | NP_001171340.1 | 9 | 1573 | |||
NM_026830.2 | NP_081106.1 | 10 | 1880 | |||
GenBank mRNA | AK147260.1 | - | 10 | 3035 | ||
AK147653.1 | - | 10 | 3035 | |||
AK154980.1 | - | 10 | 1880 | |||
AK170652.1 | - | 10 | 1912 | |||
AK171375.1 | - | 10 | 1617 | |||
AY946044.1 | - | 7 | 1264 | |||
BC080680.1 | - | 10 | 1726 | 6330949 |