Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027036.3 | NP_081312.2 | 1 | 217 | ||
GenBank mRNA | AK005588.1 | - | 1 | 216 | ||
BC048744.1 | - | 1 | 215 | |||
BC061030.1 | - | 1 | 213 | 6775221 | ||
CA465179.1 | - | 1 | 216 | |||
CN840046.1 | - | 1 | 220 |