Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039394.1 | NP_001034483.1 | 3 | 790 | ||
NM_133717.3 | NP_598478.1 | 3 | 496 | |||
GenBank mRNA | AB232640.1 | - | 3 | 555 | ||
AJ534465.1 | - | 3 | 774 | |||
AK007834.1 | - | 3 | 386 | |||
AK202533.1 | - | 1 | 76 | |||
BC010248.1 | - | 3 | 496 | 3155089 | ||
BY707837.1 | - | 3 | 383 | |||
CV782943.1 | - | 3 | 396 |