Search
Search

Fars2| Gene Symbol: Fars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039189.2 | NP_001034278.1 | 2 | 469 | ||
| NM_024274.3 | NP_077236.1 | 3 | 1102 | |||
| GenBank mRNA | AK013211.1 | - | 3 | 841 | ||
| AK020164.1 | - | 2 | 255 | |||
| AK054370.1 | - | 867 | ||||
| AK076259.1 | - | 3 | 886 | |||
| AK170986.1 | - | 3 | 885 | |||
| BC002147.1 | - | 3 | 872 | 3484842 | ||