Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027308.2 | NP_081584.1 | 4 | 829 | ||
GenBank mRNA | AK033677.1 | - | 4 | 830 | ||
AK088927.1 | - | 5 | 785 | |||
AK135657.1 | - | 635 | ||||
BC139455.1 | - | 4 | 677 | 8862477 | ||
BC139458.1 | - | 4 | 677 | 8862480 |