Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cactin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027381.2 | NP_081657.1 | 9 | 2180 | ||
GenBank mRNA | AK010963.1 | - | 6 | 989 | ||
AY934504.1 | - | 9 | 2086 | |||
BC023696.1 | - | 6 | 1100 | |||
BC150986.1 | - | 9 | 2180 | 9087899 | ||
BC151180.1 | - | 9 | 2180 | 9088069 |