siRNA Details
Gene Symbol: Ggt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027819.2 | NP_082095.2 | 4 | 1008 | ||
GenBank mRNA | BC055837.1 | - | 2 | 683 | 4225313 | |
BC058747.1 | - | 4 | 1008 | 6394797 |

Gene Symbol: Ggt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027819.2 | NP_082095.2 | 4 | 1008 | ||
GenBank mRNA | BC055837.1 | - | 2 | 683 | 4225313 | |
BC058747.1 | - | 4 | 1008 | 6394797 |