Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133733.4 | NP_598494.2 | 6 | 1065 | ||
GenBank mRNA | AB040490.1 | - | 6 | 982 | ||
AK028983.1 | - | 6 | 1035 | |||
AK033723.1 | - | 6 | 1067 | |||
AK080481.1 | - | 6 | 1021 | |||
AY259213.1 | - | 7 | 694 | |||
AY326421.1 | - | 6 | 700 | |||
BC026447.1 | - | 6 | 891 | 4459717 |
Gene Symbol: Gm16096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089373.1 | - | 428 |