Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Telo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163661.1 | NP_001157133.1 | 14 | 2243 | ||
NM_027880.2 | NP_082156.2 | 14 | 2154 | |||
GenBank mRNA | AK004582.1 | - | 14 | 2024 | ||
AK028658.1 | - | 14 | 2008 | |||
AK045321.1 | - | 7 | 1690 | |||
AK050773.1 | - | 14 | 1973 | |||
AK084830.1 | - | 14 | 2027 | |||
AK129189.1 | - | 13 | 2316 | |||
AK143827.1 | - | 14 | 2244 | |||
AK146963.1 | - | 14 | 2030 | |||
AK154727.1 | - | 14 | 2056 | |||
AK158181.1 | - | 8 | 1305 | |||
BC011077.1 | - | 14 | 2004 | 3984911 | ||
BC080713.1 | - | 13 | 2263 |