Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tysnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272091.1 | NP_001259020.1 | 2 | 1250 | ||
NM_027912.1 | NP_082188.1 | 3 | 1381 | |||
GenBank mRNA | AK005069.1 | - | 3 | 1382 | ||
AK154513.1 | - | 3 | 1364 | |||
AK185719.1 | - | 2 | 164 | |||
BC119320.1 | - | 3 | 1359 | 8734069 |