Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027963.2 | NP_082239.2 | 14 | 1841 | ||
GenBank mRNA | AK006118.1 | - | 14 | 1841 | ||
AU015948.2 | - | 1 | 358 | |||
BC118541.1 | - | 14 | 1840 | 40058756 |