Search
Search

Gpc2| Gene Symbol: Gpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172412.2 | NP_766000.1 | 3 | 470 | ||
| GenBank mRNA | AK010515.1 | - | 3 | 500 | ||
| AK049639.1 | - | 3 | 471 | |||
| AK160959.1 | - | 3 | 516 | |||
| BC056941.1 | - | 2 | 1169 | |||
| BC083180.1 | - | 3 | 477 | 5707617 | ||