Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028182.1 | NP_082458.1 | 4, 5 | 1045 | ||
GenBank mRNA | AK008803.1 | - | 4, 5 | 831 | ||
AK157396.1 | - | 5, 6 | 843 | |||
AX776203.1 | - | 3, 4 | 502 | |||
BC023784.1 | - | 4, 5 | 438 | |||
BC116682.1 | - | 4, 5 | 753 | 40106612 |