Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Cyth4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028195.3 | NP_082471.2 | 11, 12 | 1114 | ||
GenBank mRNA | AF079972.1 | - | 11, 12 | 1045 | ||
AK010908.1 | - | 1, 2 | 47 | |||
AK152306.1 | - | 11, 12 | 1020 | |||
AK152909.1 | - | 11, 12 | 1020 | |||
AK157026.1 | - | 11, 12 | 1020 | |||
AK220198.1 | - | 11, 12 | 1043 | |||
BC018505.1 | - | 10, 11 | 938 | |||
BC139054.1 | - | 11, 12 | 1087 | 8862076 | ||
BC139056.1 | - | 11, 12 | 1087 | 8862078 | ||
BC145528.1 | - | 11, 12 | 1114 |