Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178385.3 | NP_848472.2 | 1 | 964 | ||
GenBank mRNA | AK012950.1 | - | 1 | 909 | ||
AK132036.1 | - | 914 | ||||
BC019476.1 | - | 1 | 902 | 4240883 | ||
BC083164.1 | - | 1 | 896 | 5390600 |