This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
Tbkbp1
- Gene Name:
-
TBK1 binding protein 1
- Gene Aliases:
-
3110043L15Rik, ProSAPiP2, SINTBAD
- Chromosome Location:
-
Chr.11: 97136171 - 97151490 on Build GRCm38
- UniGene ID:
-
Mm.27882
- Species:
- Mouse
- Species Specific ID (Flybase ID):
-
-
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System