Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifitm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053088.2 | NP_444318.1 | 2 | 389 | ||
GenBank mRNA | AJ009781.1 | - | 2 | 389 | ||
AK003363.1 | - | 2 | 420 | |||
BC103790.1 | - | 2 | 389 | 30287396 | ||
EU380257.1 | - | 2 | 361 | |||
FJ200451.1 | - | 2 | 441 |